Thermo Scientific Chemicals

3-Amino-4-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 429860050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Amino-4-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 429860050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 429860050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3430-27-1
IUPAC Name
4-methylpyridin-3-amine
Molecular Formula
C6H8N2
InChI Key
IBKMZYWDWWIWEL-UHFFFAOYSA-N
SMILES
CC1=CC=NC=C1N
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Melting point
100°C to 108°C
HPLC
>=96.0 %
Appearance (Color)
Brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text