Copper(I) thiophene-2-carboxylate, 90%, may contain approx. 20 wt.% Copper(I) oxide
Copper(I) thiophene-2-carboxylate, 90%, may contain approx. 20 wt.% Copper(I) oxide
Copper(I) thiophene-2-carboxylate, 90%, may contain approx. 20 wt.% Copper(I) oxide
Copper(I) thiophene-2-carboxylate, 90%, may contain approx. 20 wt.% Copper(I) oxide
Thermo Scientific Chemicals

Copper(I) thiophene-2-carboxylate, 90%, may contain approx. 20 wt.% Copper(I) oxide

CAS: 68986-76-5 | C5H3CuO2S | 190.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4300100101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 430010010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS68986-76-5
IUPAC Nameλ¹-copper(1+) thiophene-2-carboxylate
Molecular FormulaC5H3CuO2S
InChI KeySFJMFSWCBVEHBA-UHFFFAOYSA-M
SMILES[Cu+].[O-]C(=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infomay contain up to 0.3 mol % Copper(I)oxide
Appearance (Color)Grey to red to brown
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis27.71 to 35.27 % (C)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
342070Certificate of AnalysisApr 04, 2022430010010
1 result displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection