Ethyl 2-butynoate, 99%, Thermo Scientific Chemicals
Ethyl 2-butynoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-butynoate, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 430030100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4341-76-8
IUPAC Nameethyl but-2-ynoate
Molecular FormulaC6H8O2
InChI KeyFCJJZKCJURDYNF-UHFFFAOYSA-N
SMILESCCOC(=O)C#CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only