Thermo Scientific Chemicals

Ethyl 2-nitrobenzoylacetate, 97%, Thermo Scientific Chemicals

Catalog number: 430040050
5 g, Each
Thermo Scientific Chemicals

Ethyl 2-nitrobenzoylacetate, 97%, Thermo Scientific Chemicals

Catalog number: 430040050
5 g, Each
Quantity
Catalog number: 430040050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
52119-39-8
IUPAC Name
ethyl 3-(2-nitrophenyl)-3-oxopropanoate
Molecular Formula
C11H11NO5
InChI Key
OWZNCVIBJQPNEF-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)C1=CC=CC=C1[N+]([O-])=O
HPLC
>=96.0 %
Appearance (Color)
Clear orange to brown
Appearance (Form)
Liquid or low melting solid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text