Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Thermo Scientific™

Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™

Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)IncludesUnit Size
4301-10001000 mL53 mm ClosureCase of 4
4301-0125Promo Image125 mL28 mm ClosureCase of 4
4301-0125PK
also known as 4301-0125
125 mLETFE ClosurePack of 1
4301-1000PK
also known as 4301-1000
1000 mLETFE ClosurePack of 1
4301-2000PK
also known as 4301-2000
2000 mLETFE ClosurePack of 1
4301-0250250 mL33 mm ClosureCase of 4
4301-0250PK
also known as 4301-0250
250 mL33 mm ClosurePack of 1
4301-0500500 mL43 mm ClosureCase of 4
4301-0500PK
also known as 4301-0500
500 mL43 mm ClosurePack of 1
4301-20002000 mL53 mm ClosureCase of 2
Catalog number 4301-1000
Price (USD)
1,286.36
Online Exclusive
1,850.00
Save 563.64 (30%)
Case of 4
Estimated availability date  Pending  
Add to cart
Capacity (Metric):
1000 mL
Includes:
53 mm Closure
Unit Size:
Case of 4
Recurring order eligible. Learn more »
Price (USD)
1,286.36
Online Exclusive
1,850.00
Save 563.64 (30%)
Case of 4
Add to cart
Ask our AI about this Product
Perform your liquid-liquid extractions with these chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer and Closure made with Tefzel™ which feature a TFE stopcock assembly. TFE stopcock assembly has excellent chemical resistance, is leakproof† when used with provided Nalgene closure and does not require lubrication — can be chemically disinfected. More transparent than our polypropylene separatory funnels. Compatible with all common lab reagents and solvents. Won't chip, crack or shatter like glass separatory funnels.
Features:
  • Break-resistant and impervious to any chemical used in a separatory funnel
  • No phthalates to contaminate analyses
  • Useful in radiological and hazardous separations
  • Autoclavable/Leakproof† when used with provided Nalgene closure
  • Stem length: 65mm

Ordering Information:

For replacement stopcock assembly for 125mL to 1L sizes, order Cat. No. 714301-1025. For replacement stopcock assembly for 2L size, order Cat. No. 714301-1050.

Note:

Funnel can be autoclaved; stopcock can be chemically sterilized. No stopcock lubrication needed.

Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Includes53 mm Closure
Quantity1/Pk.; 4/Cs.
Capacity (Metric)1000 mL
Height (Metric)371 mm
MaterialTeflon FEP
Product TypeSeparatory Funnel
Unit SizeCase of 4
Nalgene Separatory Funnels
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1317106Certificate of ComplianceDec 09, 20244301-2000
1306162Certificate of ComplianceDec 09, 20244301-1000
1317107Certificate of ComplianceDec 09, 20244301-1000
1416127Certificate of ComplianceOct 22, 20244301-1000
1408542Certificate of ComplianceJul 30, 20244301-1000
5 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection