Thermo Scientific Chemicals

1-Cyclohexenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 430180010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Cyclohexenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 430180010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 430180010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89490-05-1
IUPAC Name
(cyclohex-1-en-1-yl)boronic acid
Molecular Formula
C6H11BO2
InChI Key
XZWQKJXJNKYMAP-UHFFFAOYSA-N
SMILES
OB(O)C1=CCCCC1
Appearance (Color)
White to off-white
HPLC
>=96.0 %
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text