Thermo Scientific Chemicals

2-chloro-4-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 430190050
5 g, Each
Thermo Scientific Chemicals

2-chloro-4-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 430190050
5 g, Each
Quantity
Catalog number: 430190050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
313545-72-1
IUPAC Name
(2-chloro-4-fluorophenyl)boronic acid
Molecular Formula
C6H5BClFO2
InChI Key
XOFNMNLYGPKKOV-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(F)C=C1Cl
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text