2-Chloro-3-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals
2-Chloro-3-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-3-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 430370010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS871329-52-1
IUPAC Name(2-chloro-3-fluorophenyl)boronic acid
Molecular FormulaC6H5BClFO2
InChI KeyTYOGIUGNZOBBHE-UHFFFAOYSA-N
SMILESOB(O)C1=C(Cl)C(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point113°C to 117°C
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only