Thermo Scientific Chemicals

2-Methylpyridine-3-boronic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 430390010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methylpyridine-3-boronic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 430390010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 430390010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1072952-34-1
IUPAC Name
(2-methylpyridin-3-yl)boronic acid hydrochloride
Molecular Formula
C6H9BClNO2
InChI Key
OAMCCRLFDPJOPN-UHFFFAOYSA-N
SMILES
Cl.CC1=C(C=CC=N1)B(O)O
Infrared spectrum
Conforms
Appearance (Color)
Off-white
HPLC
>=96.0 %
Appearance (Form)
Crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text