Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 430570010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1168135-03-2
IUPAC Name5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
Molecular FormulaC12H15BN2O2S
InChI KeyKISHNZJGTMYYKH-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC2=NSN=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point83°C to 87°C
Appearance (Color)Brown
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only