Thermo Scientific Chemicals

2-Nitrophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 430590010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Nitrophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 430590010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 430590010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5570-19-4
IUPAC Name
(2-nitrophenyl)boronic acid
Molecular Formula
C6H6BNO4
InChI Key
SFUIGUOONHIVLG-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC=C1[N+]([O-])=O
HPLC
>=96.0 %
Appearance (Color)
Brown
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text