Thermo Scientific Chemicals

2-chloropyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 430660050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-chloropyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 430660050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 430660050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
452972-11-1
IUPAC Name
2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Formula
C11H15BClNO2
InChI Key
XFZFMAUUZHBQSS-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=CN=C1Cl
GC
>=96.0 %
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or low melting solid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text