Thermo Scientific Chemicals

2-Formylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 430670010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Formylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 430670010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 430670010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
380151-85-9
IUPAC Name
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular Formula
C13H17BO3
InChI Key
SLJMPQLPRHIAOM-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=CC=C1C=O
Appearance (Color)
Clear colorless to dark yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text