1-Pentene, 97%, AcroSeal™
1-Pentene, 97%, AcroSeal™
1-Pentene, 97%, AcroSeal™
1-Pentene, 97%, AcroSeal™
1-Pentene, 97%, AcroSeal™
Thermo Scientific Chemicals

1-Pentene, 97%, AcroSeal™

CAS: 109-67-1 | C5H10 | 70.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
430731000100 mLAcroSeal™ Glass Bottle
Catalog number 430731000
Price (USD)
139.00
Each
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
139.00
Each
Add to cart
Chemical Identifiers
CAS109-67-1
IUPAC Namepent-1-ene
Molecular FormulaC5H10
InChI KeyYWAKXRMUMFPDSH-UHFFFAOYSA-N
SMILESCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.3704 to 1.3724 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link