Applied Biosystems™

StockMarks™ for Dogs Genotyping Kit, canine

Catalog number: 4307481
Have Questions?
Applied Biosystems™

StockMarks™ for Dogs Genotyping Kit, canine

Catalog number: 4307481
Have Questions?
Catalog Number
4307481
Unit Size
100 assays
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4307481100 assays
-
4349041100 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The StockMarks™ for Dogs Canine Genotyping Kit is part of a complete solution for animal parentage typing and genotyping employing short tandem repeats (STRs), also referred to as microsatellites. Microsatellite markers are evenly distributed across genome. They are highly polymorphic and they can be identified within DNA samples using PCR.

The StockMarks™ kits employ a proprietary PCR process for amplification of the microsatellite markers using fluorescent dyelabeled primers. The StockMarks™ for Cattle and StockMarks™ for Horses kits amplify the microsatellite markers recommended by the International Society of Animal Genetics (ISAG), while the StockMarks™ for Dogs kit amplifies the microsatellite markers recommended by the American Kennel Club (AKC).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Capillary Electrophoresis
Format
Kit
Sample Type
DNA
For Use With (Equipment)
310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, GeneAmp 9600, GeneAmp 9700
For Use With (Application)
Sequencing
Hot Start
Built-In Hot Start
Label or Dye
FAM, JOE, NED
No. of Reactions
100 Assays
PCR Method
Multiplex PCR
Polymerase
AmpliTaq™ Gold DNA Polymerase
Product Line
StockMarks™
Product Type
Genotyping Kit
Quantity
100 Assays
Shipping Condition
Dry Ice
Species
Dog/Canine
Target
Canine Genotyping and Parentage Testing
Target Loci
FHC 2054, FHC 2010, PEZ 3, FHC 2079, PEZ 20, PEZ 1, PEZ 12, PEZ 8, PEZ 5, PEZ 6
Technique
STR (Short Tandem Repeat) Analysis
Unit Size
100 assays

Contents & Storage

This kit contains labeled and unlabeled primer mix, control DNA, AmliTaq Gold DNA polymerase, dNTPs and buffers. There are enough reagents to perform 100 tests.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text