Succinyl chloride, ca. 95%, AcroSeal™
Succinyl chloride, ca. 95%, AcroSeal™
Succinyl chloride, ca. 95%, AcroSeal™
Succinyl chloride, ca. 95%, AcroSeal™
Succinyl chloride, ca. 95%, AcroSeal™
Thermo Scientific Chemicals

Succinyl chloride, ca. 95%, AcroSeal™

CAS: 543-20-4 | C4H4Cl2O2 | 154.98 g/mol
Have Questions?
Catalog NumberQuantityPackaging
430801000100 mLAcroSeal™ Glass Bottle
Catalog number 430801000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS543-20-4
IUPAC Namebutanedioyl dichloride
Molecular FormulaC4H4Cl2O2
InChI KeyIRXBNHGNHKNOJI-UHFFFAOYSA-N
SMILESClC(=O)CCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light brown
Refractive index1.4670 to 1.4720 (20°C, 589 nm)
Appearance (Form)Liquid
Titration RCOCl>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link