Thermo Scientific Chemicals

Dimethyl sulfate, 99+%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 430830010
1 L, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Dimethyl sulfate, 99+%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 430830010
1 L, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 430830010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-78-1
IUPAC Name
dimethyl sulfate
Molecular Formula
C2H6O4S
InChI Key
VAYGXNSJCAHWJZ-UHFFFAOYSA-N
SMILES
COS(=O)(=O)OC
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Oily liquid
Infrared spectrum
Conforms
Refractive index
1.3860 to 1.3880 (20°C, 589 nm)
Acidity
=<0.2 % (as Sulfuric acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text