Dimethyl sulfate, 99+%, AcroSeal™
Dimethyl sulfate, 99+%, AcroSeal™
Dimethyl sulfate, 99+%, AcroSeal™
Dimethyl sulfate, 99+%, AcroSeal™
Dimethyl sulfate, 99+%, AcroSeal™
Thermo Scientific Chemicals

Dimethyl sulfate, 99+%, AcroSeal™

CAS: 77-78-1 | C2H6O4S | 126.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 430831000
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Add to cart
Chemical Identifiers
CAS77-78-1
IUPAC Namedimethyl sulfate
Molecular FormulaC2H6O4S
InChI KeyVAYGXNSJCAHWJZ-UHFFFAOYSA-N
SMILESCOS(=O)(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow to light brown
Appearance (Form)Oily liquid
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
Titration99.0 to 101.0 % (Hydrolysis)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0466877Certificate of AnalysisMay 17, 2025430830010, 430831000
A0462037Certificate of AnalysisMay 17, 2025430830010, 430831000
A0466877Certificate of AnalysisOct 15, 2024430830010, 430831000
A0462037Certificate of AnalysisApr 15, 2024430830010, 430831000
A0460169Certificate of AnalysisFeb 12, 2024430830010, 430831000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection