Methyl chloroformate, 99%, AcroSeal™, Thermo Scientific Chemicals
Methyl chloroformate, 99%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl chloroformate, 99%, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 430841000
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.60 
Save 5.95 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.60 
Save 5.95 (15%)
Add to cart
Methyl chloroformate, 99%, AcroSeal™, Thermo Scientific Chemicals
Catalog number430841000
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.60 
Save 5.95 (15%)
-
Add to cart
Chemical Identifiers
CAS79-22-1
IUPAC Namemethyl carbonochloridate
Molecular FormulaC2H3ClO2
InChI KeyXMJHPCRAQCTCFT-UHFFFAOYSA-N
SMILESCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only