(±)-Propylene oxide, 99.5%, extra pure, AcroSeal™, Thermo Scientific Chemicals
(±)-Propylene oxide, 99.5%, extra pure, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(±)-Propylene oxide, 99.5%, extra pure, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 430861000
Price (USD)/ Each
23.65
Online exclusive
26.00 
Save 2.35 (9%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
23.65
Online exclusive
26.00 
Save 2.35 (9%)
Add to cart
(±)-Propylene oxide, 99.5%, extra pure, AcroSeal™, Thermo Scientific Chemicals
Catalog number430861000
Price (USD)/ Each
23.65
Online exclusive
26.00 
Save 2.35 (9%)
-
Add to cart
Chemical Identifiers
CAS75-56-9
IUPAC Name2-methyloxirane
Molecular FormulaC3H6O
InChI KeyGOOHAUXETOMSMM-UHFFFAOYNA-N
SMILESCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Aldehyde (C3H6O)=<0.01 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only