Thermo Scientific Chemicals

Dichlorophenylsilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 430898000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Dichlorophenylsilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 430898000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 430898000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-10-4
IUPAC Name
dichlorodiphenylsilane
Molecular Formula
C12H10Cl2Si
InChI Key
OSXYHAQZDCICNX-UHFFFAOYSA-N
SMILES
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to light brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5765 to 1.5815 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text