Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ Optical 384-Well Reaction Plate with Barcode

Catalog number: 4309849
Have Questions?
Applied Biosystems™

MicroAmp™ Optical 384-Well Reaction Plate with Barcode

Catalog number: 4309849
Have Questions?
Catalog Number
4309849
Unit Size
50 plates
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 plates
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
430984950 plates
-
4326270500 plates
-
43438141,000 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ MicroAmp™ Optical 384-Well Reaction Plate with Barcode is designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 384-well format, is designed to work with Applied Biosystems™ 384-well Real-Time PCR systems and thermal cyclers.

• Frosted to minimize interfering fluorescence from wells of cycling block
• Screened to eliminate auto-fluorescent plates
• Unique design provides a barrier to ambient air to help ensure well-to-well temperature uniformity
• Each reaction plate includes a unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors

Oil-Free PCR Amplification
MicroAmp™ disposables are optimized to provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification. Proprietary MicroAmp™ plates are essential for high-performance PCR and help ensure the greatest temperature and amplification uniformity for your samples.

Specifications

Barcode
Includes Barcode
Certifications/Compliance
Certified DNA/RNase Free
Packaging
50 Plates
Product Line
MicroAmp
Color
Optical
Product Type
Microplate
For Use With (Equipment)
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style
Skirted
Material
Polypropylene
Volume (Metric) Well
40 μL
No. of Wells
384
Includes
50 plates
Reaction Speed
Fast or Standard
Shipping Condition
Room Temperature

Contents & Storage

Contains 50 plates.

May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text