Thermo Scientific Chemicals

Isoquinoline-8-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 431050010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Isoquinoline-8-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 431050010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 431050010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61563-43-7
IUPAC Name
isoquinoline-8-carboxylic acid
Molecular Formula
C10H7NO2
InChI Key
VMNZQPRIUSJCOH-UHFFFAOYSA-N
SMILES
OC(=O)C1=C2C=NC=CC2=CC=C1
Appearance (Form)
Crystalline powder
Appearance (Color)
White to light brown
Infrared spectrum
Conforms
HPLC
>=96.0 %
Melting point
263°C to 267°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text