Hi-Di™ Formamide
Hi-Di™ Formamide
Applied Biosystems™

Hi-Di™ Formamide

This is highly deionized formamide used to resuspend samples before electrokinetic injection on capillary electrophoresis systems.For Research Use Only. NotRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
431132025 mL
44407534 x 5 mL
Catalog number 4311320
Price (USD)
61.00
Each
Add to cart
Quantity:
25 mL
Price (USD)
61.00
Each
Add to cart
This is highly deionized formamide used to resuspend samples before electrokinetic injection on capillary electrophoresis systems.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
FormLiquid
Product LineHigh-Di™
Product TypeFormamide
Quantity25 mL
Reagent TypeFormamide
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store at -15°C to -25°C. Avoid multiple freeze-thaw cycles.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link