Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ Splash-Free 96-Well Base

Catalog number: 4312063
Have Questions?
Applied Biosystems™

MicroAmp™ Splash-Free 96-Well Base

Catalog number: 4312063
Have Questions?
Catalog Number
4312063
Unit Size
10 bases
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
431206310 bases
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems™ MicroAmp™ Bases are designed to hold MicroAmp™ Reaction Tubes in MicroAmp™ Tray/Retainer Sets or 96-Well Reaction Plates for convenient pre-PCR and post-PCR sample processing.

• Perfect for centrifugation

Uniform Results

Use with instruments that have heated sample covers to eliminate condensation and the need for a mineral oil overlay. Proprietary MicroAmp™ Reaction Tubes are essential for high-performance PCR. They ensure the greatest temperature and amplification uniformity for your samples.

Note:
See user's manual or package insert for limited label license and trademark information.

Specifications

Certifications/Compliance
Certified DNA/RNase Free
Product Line
MicroAmp
Color
Black
Product Type
96-Well Base
For Use With (Equipment)
7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOne™, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, 7500 Fast Dx System, 2720 Thermal Cycler, ViiA™ 7 Fast System, QuantStudio™ Dx
For Use With (Application)
PCR
Plate Compatibility
96-Well Plates
Quantity
10 Bases
Shipping Condition
Room Temperature

Contents & Storage

Contains 10 MicroAmp™ Splash Free 96-Well Bases. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text