Applied Biosystems™

TaqMan™ Control Genomic DNA (human)

Catalog number: 4312660
Have Questions?
Applied Biosystems™

TaqMan™ Control Genomic DNA (human)

Catalog number: 4312660
Have Questions?
Catalog Number
4312660
Unit Size
2 tubes
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43126602 tubes
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Human Genomic DNA Control provided at a convenient PCR-ready concentration.

Applied Biosystems™ TaqMan™ Control Genomic DNA (Human) is conveniently packaged in a PCR-ready concentration for use as a control template in PCR reactions. The Human gDNA (male) is provided in two tubes (100 μL each) at a concentration of 10 ng⁄μL. TaqMan™ Control Genomic Human DNA is also included in several Applied Biosystems™ real-time PCR kits.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Frozen
Species
Human
Shipping Condition
Dry Ice
Concentration
10 ng/μL
Label or Dye
Unlabeled
Quantity
2 Tubes
Product Line
TaqMan™

Contents & Storage

Human gDNA (Male) packaged in two tubes (100μL each). Store at -15 to 25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text