trans-2-(4-chlorophenyl)vinylboronic acid, 97%, Thermo Scientific Chemicals
trans-2-(4-chlorophenyl)vinylboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-(4-chlorophenyl)vinylboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number 431340100
Price (USD)/ Each
605.65
Online exclusive
673.00 
Save 67.35 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
605.65
Online exclusive
673.00 
Save 67.35 (10%)
Add to cart
trans-2-(4-chlorophenyl)vinylboronic acid, 97%, Thermo Scientific Chemicals
Catalog number431340100
Price (USD)/ Each
605.65
Online exclusive
673.00 
Save 67.35 (10%)
-
Add to cart
Chemical Identifiers
CAS154230-29-2
IUPAC Name[(1E)-2-(4-chlorophenyl)ethenyl]boronic acid
Molecular FormulaC8H8BClO2
InChI KeyHWSDRAPTZRYXHN-AATRIKPKSA-N
SMILESOB(O)\C=C\C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only