MicroAmp™ Optical Adhesive Film Kit
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ Optical Adhesive Film Kit
Applied Biosystems™
MicroAmp™ Optical Adhesive Film Kit
The Applied Biosystems™ MicroAmp™ Optical Adhesive Film Kit includes the following products:• MicroAmp™ Optical Adhesive Film reduces the chance ofRead more
Have Questions?
Catalog NumberQuantity
43136631 kit
Catalog number 4313663
Price (USD)
102.65
Online Exclusive
107.00
Save 4.35 (4%)
Each
-
Add to cart
Quantity:
1 kit
Price (USD)
102.65
Online Exclusive
107.00
Save 4.35 (4%)
Each
Add to cart
Ask our AI about this Product
The Applied Biosystems™ MicroAmp™ Optical Adhesive Film Kit includes the following products:
• MicroAmp™ Optical Adhesive Film reduces the chance of well-to-well contamination and sample evaporation when applied to a microplate.
• MicroAmp™ Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films
• MicroAmp™ Adhesive Film Applicator is used to form a tight seal between a microplate and an adhesive film.
MicroAmp™ Optical Adhesive Film reduces the chance of well-to-well contamination and sample evaporation when applied to a microplate.
• Pressure-sensitive adhesive provides a tight seal over every well
• Will not interfere with sample reading
• Press-N-Seal: Align properly to the plate, then press to seal
• Easy to apply-does not stick to your glove
Prevent Loss of Precious Samples
Use this optically-clear adhesive film to seal the samples into the wells of a microplate. This will reduce the possibility of cross-contamination between sample wells and help ensure consistent real-time PCR data.
Easy to Use
Simply peel the backing away from the adhesive film and apply film firmly across the top of your microplate. Stack microplates and read samples as you would normally except with less chance of sample contamination. Designed to fit on MicroAmp™ Optical 96-well Reaction Plates.
MicroAmp™ Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films.
Use the MicroAmp™ Adhesive Film Applicator to form a tight seal between a microplate and the adhesive film.
Form a Tight Seal
The MicroAmp™ Adhesive Film Applicator enables you to ergonomically push an adhesive film down upon a microplate to form a tight seal and minimize air bubbles. This tight seal will prevent well-to-well contamination and sample loss.
Quick and Easy
Use the MicroAmp™ Adhesive Film Applicator to speed the process of sealing multiple microplates. This tool is designed according to the microplate dimensions so you can quickly seal a microplate. The rugged proprietary plastic endures constant use so you can complete large screening applications.
Note: See user's manual or package insert for limited label license, and trademark information.
Specifications
For Use With (Equipment)2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7900HT System, GeneAmp 9700, QuantStudio™, ViiA™ 7 Dx System, ViiA™ 7 System, QuantStudio™ Dx
Product TypeFilm, Seal, or Applicator
Quantity1 kit
Shipping ConditionRoom Temperature
AdhesivePressure Sensitive
ColorOptical
MaterialPolyester
No. of Wells96
Plate Compatibility96-well Plate
Product LineMicroAmp
Unit SizeEach
Contents & Storage
Includes: MicroAmp™ Optical Adhesive Covers (quantity 20), Applicator (quantity 1), MicroAmp™ Optical Cover Compression Pad (quantity 1)
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
How do I correctly apply MicroAmp Optical Adhesive Covers to an optical 96- or 384-well plate?
Please follow the instructions below for the proper application of MicroAmp Optical Adhesive Covers (Cat. Nos. 4311971, 4313663 and 4360954).
The MicroAmp Optical Adhesive Cover uses a pressure-sensitive adhesive backing to adhere the cover to the optical 96- or 384-well plate. It is imperative that you use enough force to activate the pressure-sensitive adhesive to prevent evaporation from the wells.
Please note: Users should not expect the Optical Adhesive Cover to be very sticky to the touch on the side that adheres to the optical plate.
The correct way to apply the cover is as follows:
1. Apply firm pressure when using the Adhesive Seal Applicator (Cat. No. 4333183) to press the cover onto the plate.
2. Run the applicator firmly from left to right several times. Run the applicator up and down several times.
3. Run the edge of the applicator around the border (following the red line outside the wells) of the cover.
4. Repeat the left to right and up and down motions to assure a good final seal.
5. Inspect each well to make sure that each well edges are imprinted on the optical adhesive cover.
Please Note: Using the Applicator's edge to seal between the wells is NOT recommended. It may actually pull the seal off the wells.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?
No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection