Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films

Catalog number: 4314320
Have Questions?
Applied Biosystems™

MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films

Catalog number: 4314320
Have Questions?
Catalog Number
4314320
Unit Size
100 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4314320100 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate is optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems™ 96-Well Real-Time PCR systems and thermal cyclers.

• Frosted to minimize interfering fluorescence from wells of cycling block
• Screened to eliminate auto-fluorescent plates
• Unique design provides a barrier to ambient air to help insure well-to-well temperature uniformity
• Each reaction plate includes a unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors

The included MicroAmp™ Optical Adhesive Films reduce the chance of well-to-well contamination and sample evaporation when applied to the microplates.

• Pressure-sensitive adhesive provides a tight seal over every well
• Will not interfere with sample reading
• Confirm correct alignment to plate, then press to seal
• Easy to apply; does not stick to your glove

Oil-Free PCR Amplification
MicroAmp™ disposables are optimized to provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification. Proprietary MicroAmp™ plates are essential for high-performance PCR and help ensure the greatest temperature and amplification uniformity for your samples.

Specifications

Color
Optical
Certifications/Compliance
Certified DNA/RNase Free
Material
Polypropylene
No. of Wells
96
Format
96-well plate
For Use With (Equipment)
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Product Type
Plate/Adhesive Film/Barcode
Product Line
MicroAmp
Adhesive
Pressure Sensitive
Plate Compatibility
96-well Plate

Contents & Storage

Contains:
5 packages of 20 plates each (equivalent to 5 × catalog # 4306737)
1 package of 100 Optical Adhesive Films (equivalent to catalog # 4311971)

Store at room temperature. With proper storage, shelf-life is 2 years.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text