Thermo Scientific Chemicals

Ethyl N-BOC-4-piperidinecarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 431500050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl N-BOC-4-piperidinecarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 431500050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 431500050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142851-03-4
IUPAC Name
1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate
Molecular Formula
C13H23NO4
InChI Key
MYHJCTUTPIKNAT-UHFFFAOYSA-N
SMILES
CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text