Ethyl N-BOC-4-piperidinecarboxylate, 97%
Ethyl N-BOC-4-piperidinecarboxylate, 97%
Ethyl N-BOC-4-piperidinecarboxylate, 97%
Ethyl N-BOC-4-piperidinecarboxylate, 97%
Thermo Scientific Chemicals

Ethyl N-BOC-4-piperidinecarboxylate, 97%

CAS: 142851-03-4 | C13H23NO4 | 257.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 431500250
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS142851-03-4
IUPAC Name1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate
Molecular FormulaC13H23NO4
InChI KeyMYHJCTUTPIKNAT-UHFFFAOYSA-N
SMILESCCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link