Thermo Scientific Chemicals

Oxolinic acid, 98%, Thermo Scientific Chemicals

Catalog number: 431510250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Oxolinic acid, 98%, Thermo Scientific Chemicals

Catalog number: 431510250
25 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 431510250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14698-29-4
Appearance (Form)
Crystalline powder
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Melting point
311°C to 314°C (Dec)
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text