Thermo Scientific Chemicals

3,4-Dichlorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431570500
50 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

3,4-Dichlorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431570500
50 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 431570500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79175-35-2
IUPAC Name
bromo(3,4-dichlorophenyl)magnesium
Molecular Formula
C6H3BrCl2Mg
InChI Key
IRROSODEQHIZEV-UHFFFAOYSA-M
SMILES
ClC1=CC=C([Mg]Br)C=C1Cl
Appearance (Form)
Liquid
Appearance (Color)
Yellow to brown
Titration Grignard
0.40 to 0.50 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text