Thermo Scientific Chemicals

Cesium methanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 431580050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Cesium methanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 431580050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 431580050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2550-61-0
IUPAC Name
caesium(1+) methanesulfonate
Molecular Formula
CH3CsO3S
InChI Key
DJKJXRLREATOMF-UHFFFAOYSA-M
SMILES
[Cs+].CS([O-])(=O)=O
Appearance (Form)
Powder
Appearance (Color)
White
Infrared spectrum
Conforms
Solubility
(5 % in water) Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text