Thermo Scientific Chemicals

kappa-Carrageenan, Thermo Scientific Chemicals

Catalog number: 431591000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

kappa-Carrageenan, Thermo Scientific Chemicals

Catalog number: 431591000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 431591000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11114-20-8
IUPAC Name
(2R,3R,4R,5R,6S)-6-{[(1R,3R,4R,5R,8S)-3-{[(2S,3R,4R,5S,6R)-2-{[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3-hydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl sulfate
Molecular Formula
C24H36O25S2
InChI Key
ZNOZWUKQPJXOIG-XSBHQQIPSA-L
SMILES
OC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](O)[C@@H](O[C@@H]2[C@@H](O)[C@H](O[C@H]4[C@H]5CO[C@@H]4[C@@H](O)[C@@H](O)O5)O[C@H](CO)[C@@H]2OS([O-])(=O)=O)O3)[C@H](O)[C@@H](O)[C@H]1OS([O-])(=O)=O
Ash
=<40 %
Infrared spectrum
Conforms
Appearance (Color)
White to off-white to yellow to brown
Appearance (Form)
Powder
Loss on drying
=<12 % (1 g, 105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text