Applied Biosystems™

Plate Septa, 96 well

Catalog number: 4315933
Have Questions?
Applied Biosystems™

Plate Septa, 96 well

Catalog number: 4315933
Have Questions?
Catalog Number
4315933
Unit Size
20 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
431593320 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
These are septa for use with the 3730, 3730xl, 3130xl, and 3130, instruments.

Use with:
310 Running Buffer (10X), 3100/3100-Avant™ Genetic Analyzer Autosampler Plate Kit, 96-well, 3130/3100-Avant™ Genetic Analyzer Capillary Array (36 cm), 3130xl/3100 Genetic Analyzer Capillary Array (36 cm), 3130xl/3100 Genetic Analyzer Capillary Array (50 cm), 3730 Running Buffer (10X), 5.0 mL Glass Syringe (polymer reserve syringe), 96-Well Plate Base, 96-Well Plate Retainer, Hi-Di™ Formamide, MicroAmp™ Optical 96-Well Reaction Plate, MicroAmp™ Optical 96-Well Reaction Plate with Barcode, POP-4™ Polymer for 3100/3100-Avant™ Genetic Analyzers, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-7™ Polymer for 3130/3130xl Genetic Analyzers, POP-7™ Polymer for 3130/3130xl Genetic Analyzers, Reservoir Septa
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
96-well Plate
For Use With (Equipment)
3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3730 DNA Analyzer, 3130 Genetic Analyzer
Product Type
Septa Plate
Shipping Condition
Room Temperature

Contents & Storage

Each package contains 20 septa, sufficient for 1,920 samples.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text