4-Isopropylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
4-Isopropylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isopropylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 431630500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS18620-03-6
IUPAC Namebromo[4-(propan-2-yl)phenyl]magnesium
Molecular FormulaC9H11BrMg
InChI KeyVLHDTCQJXYKEJT-UHFFFAOYSA-M
SMILESCC(C)C1=CC=C([Mg]Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration Grignard0.40 to 0.60 M
Appearance (Color)Yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only