MicroAmp™ Optical 8-Tube Strip, 0.2 mL
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ Optical 8-Tube Strip, 0.2 mL
Applied Biosystems™

MicroAmp™ Optical 8-Tube Strip, 0.2 mL

The Applied Biosystems™ MicroAmp™ Optical 8-Tube Strip is optimally designed to fit snugly in our Applied Biosystems™ Real-Time PCR SystemsRead more
Have Questions?
Catalog NumberQuantity
4316567125 Strips
Catalog number 4316567
Price (USD)
159.65
Online Exclusive
168.00
Save 8.35 (5%)
Each
-
Add to cart
Quantity:
125 Strips
Price (USD)
159.65
Online Exclusive
168.00
Save 8.35 (5%)
Each
Add to cart
Ask our AI about this Product
The Applied Biosystems™ MicroAmp™ Optical 8-Tube Strip is optimally designed to fit snugly in our Applied Biosystems™ Real-Time PCR Systems or Thermal Cyclers, maximizing thermal conductivity and providing precise control of cycling temperatures. Requires a tray or retainer tray specific to your instrument.

Note: See user's manual or package insert for limited label license, and trademark information.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
For Use With (Equipment)Thermal Cyclers or Real-Time PCR Systems
Format(125) 8-Tube Strips
Product LineMicroAmp™ 
Quantity125 Strips
Reaction SpeedStandard
Shipping ConditionRoom Temperature
Volume (Metric) Working0.2 mL
Closure TypeNot Included
ColorOptical
MaterialPolypropylene
Product TypeOptical 8-Tube Strip
Volume (Metric)0.25 mL
Unit SizeEach
Contents & Storage
Contents: 1000 Tubes in strips of 8
Storage: Room temperature
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I run strip tubes on the ViiA 7 instrument?

Yes. Make sure to use the appropriate matching parts. For the 96-well standard block, use the standard 96-well plate adapter (Cat. No. 4459845), the retainer tray for (0.2 mL) tubes, and Optical 8-Tube Strips and Caps (Cat. No. 4316567 and 4323032). For the 96-well Fast block, use the Fast 96-well plate adapter (Cat. No. 4462078), the retainer tray (Cat. No. 4379983), and Fast (0.1 mL) Optical 8-tube Caps and Strips (Cat. Nos. 4323032 and 4358293).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What is the difference between the MicroAmp Optical Tubes (P/N801-0933) and the ABI PRISM Optical Tubes (P/N 4316567)? Is there any difference in the quality of the plastic used for the two different types?

There is no difference between these tubes.The MicroAmp Optical Tubes (P/N N801-0933) and the ABI PRISM Optical Tubes (P/N 4316567) are both made of a high-grade polypropylene. The tubes are frosted on the bottom and the caps provide for enhanced light transfer. Using non-optical tubes can give rise to significant attenuations in signal and therefore will lead to potential decreases in sensitivity in real-time assays.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What does the SeqStudio Starter Kit (PN A35000) contain?

The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link