[2-(4-Morpholinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, Thermo Scientific Chemicals
[2-(4-Morpholinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

[2-(4-Morpholinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 431750500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS480424-77-9
IUPAC Name4-{[2-(bromomagnesio)phenyl]methyl}morpholine
Molecular FormulaC11H14BrMgNO
InChI KeyRSBWIKCTZYDPJQ-UHFFFAOYSA-M
SMILESBr[Mg]C1=C(CN2CCOCC2)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear yellow
Titration Grignard0.15 to 0.35 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only