Thermo Scientific Chemicals

3-Methoxybenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431820500
50 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

3-Methoxybenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431820500
50 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 431820500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26905-40-8
IUPAC Name
chloro[(3-methoxyphenyl)methyl]magnesium
Molecular Formula
C8H9ClMgO
InChI Key
ZPCUEMLXJGELGI-UHFFFAOYSA-M
SMILES
COC1=CC=CC(C[Mg]Cl)=C1
Titration Grignard
0.15 to 0.35 M
Appearance (Form)
Solution
Appearance (Color)
Clear yellow to grey to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text