Thermo Scientific Chemicals

5-Fluoro-2-methoxyphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431880500
50 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

5-Fluoro-2-methoxyphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431880500
50 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 431880500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
188132-02-7
IUPAC Name
bromo(5-fluoro-2-methoxyphenyl)magnesium
Molecular Formula
C7H6BrFMgO
InChI Key
ZYVBZHZWABCRCW-UHFFFAOYSA-M
SMILES
COC1=CC=C(F)C=C1[Mg]Br
Titration Grignard
0.40 to 0.60 M
Appearance (Color)
Clear yellow to brown
Appearance (Form)
Solution
Appearance
May develop some turbidity or precipitate

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text