3,4-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
3,4-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 431910500
Price (USD)/ Each
326.00
-
Add to cart
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
326.00
Add to cart
3,4-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Catalog number431910500
Price (USD)/ Each
326.00
-
Add to cart
Chemical Identifiers
IUPAC Namebromo(3,4-dimethylphenyl)magnesium
Molecular FormulaC8H9BrMg
InChI KeyVMCHYKKMHINIHC-UHFFFAOYSA-M
SMILESCC1=CC=C([Mg]Br)C=C1C
Molecular Weight (g/mol)209.37
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.40 to 0.60 M
Appearance (Form)Solution
Appearance (Color)Yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only