Eukaryotic 18S rRNA Endogenous Control (VIC™/MGB probe, primer limited)
Eukaryotic 18S rRNA Endogenous Control (VIC™/MGB probe, primer limited)
Applied Biosystems™

Eukaryotic 18S rRNA Endogenous Control (VIC™/MGB probe, primer limited)

Green features
The Applied Biosystems™ Eukaryotic 18S rRNA Endogenous Control (VIC™ ⁄ MGB Probe, Primer Limited) is intended as an endogenous control.Read more
Have Questions?
Catalog number 4319413E
Price (USD)/ 2500 reactions
853.65
Online exclusive
952.00 
Save 98.35 (10%)
-
Add to cart
Price (USD)/ 2500 reactions
853.65
Online exclusive
952.00 
Save 98.35 (10%)
Add to cart
Eukaryotic 18S rRNA Endogenous Control (VIC™/MGB probe, primer limited)
Catalog number4319413E
Price (USD)/ 2500 reactions
853.65
Online exclusive
952.00 
Save 98.35 (10%)
-
Add to cart
The Applied Biosystems™ Eukaryotic 18S rRNA Endogenous Control (VIC™ ⁄ MGB Probe, Primer Limited) is intended as an endogenous control. It allows relative gene expression quantification in cDNA samples when used with other gene expression assays. Probe is labeled with VIC™ dye - MGB and the primers are limited. Can be used for multiplex or singleplex PCR reactions. Endogenous control is to be used with Inventoried and Non-Inventoried TaqMan Gene Expression Assays, Custom Gene Expression Assays, and⁄or Custom TaqMan™ Primers and Probes.

Assay Details:

Gene Symbol: 18S
RefSeq: X03205.1
Probe Exon Location: NA
Amplicon Size: 187
Corresponding TaqMan Assay ID: Hs99999901_s1

TaqMan™ Endogenous Controls

Eliminate months of assay design, formulation, and testing by using TaqMan™ Endogenous Controls as your controls to quantify gene expression. This convenient collection of pre-designed probe and primer sets enables you to normalize the amount of sample RNA or DNA in a reaction.

Complete Solution for Quantitative Gene Expression

Having a hard time deciding what controls to use to quantify gene expression — even with detailed information on biological systems? Now, with TaqMan™ Endogenous Controls, you can avoid all the trial-and-error of selecting controls for most common human, mouse, rat, and eukaryotic genes.

Simple to Use

All components of the TaqMan™ Endogenous Controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan™ Universal PCR Master Mix (with or without AmpErase™ UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.

Flexible Offering

We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
• A FAM™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (900nM each)
• A VIC™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)
• A VIC™ dye-labeled TAMRA™ probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)

Choosing the Right Endogenous Control

Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.

Multiplex vs. Singleplex PCR

All TaqMan™ Endogenous Controls that contain probes labeled with the VIC™ reporter dye are primer limited. This allows multiplexing of TaqMan™ Endogenous Controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan™ Endogenous Controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing .

For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberX03205.1
Amplicon Size187
Detection MethodPrimer-probe
FormLiquid
FormatTube
PCR Method2-step RT-qPCR
Gene Symbol18S
Green FeaturesSustainable packaging
Label or DyeVIC
Product LineTaqMan™, VIC™
Shipping ConditionRoom Temperature
SpeciesEukaryotic, Rat, Mouse, Human
Target18S rRNA
For Use With (Application)Gene Expression
Concentration20X
No. of Reactions2500 Reactions
Unit Size2500 reactions
Contents & Storage
Control Primer-Probe Set(s)