Thermo Scientific Chemicals

3-Chloro-4-fluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431990500
50 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

3-Chloro-4-fluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 431990500
50 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 431990500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
413589-34-1
IUPAC Name
bromo(3-chloro-4-fluorophenyl)magnesium
Molecular Formula
C6H3BrClFMg
InChI Key
XRNCDMQVKMYTAM-UHFFFAOYSA-M
SMILES
FC1=CC=C([Mg]Br)C=C1Cl
Appearance (Color)
Yellow to brown
Appearance (Form)
Solution
Titration Grignard
0.40 to 0.60 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text