Thermo Scientific Chemicals

N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ, Thermo Scientific Chemicals

Catalog number: 432025000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ, Thermo Scientific Chemicals

Catalog number: 432025000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 432025000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2680-03-7
IUPAC Name
N,N-dimethylprop-2-enamide
Molecular Formula
C5H9NO
InChI Key
YLGYACDQVQQZSW-UHFFFAOYSA-N
SMILES
CN(C)C(=O)C=C
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4730 to 1.4750 (20°C, 589 nm)
Infrared spectrum
Conforms
Stabilizer
Approx. 500 ppm
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text