Thermo Scientific Chemicals

(R)-1-BOC-3-methylpiperazine, 99%, Thermo Scientific Chemicals

Catalog number: 432050010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-1-BOC-3-methylpiperazine, 99%, Thermo Scientific Chemicals

Catalog number: 432050010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 432050010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
163765-44-4
IUPAC Name
tert-butyl (3R)-3-methylpiperazine-1-carboxylate
Molecular Formula
C10H20N2O2
InChI Key
FMLPQHJYUZTHQS-MRVPVSSYSA-N
SMILES
C[C@@H]1CN(CCN1)C(=O)OC(C)(C)C
Appearance (Form)
Crystalline powder
GC
>=98.5 %
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
43°C to 47°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text