Thermo Scientific Chemicals

Tetrabutylammonium chloride hydrate, 96%, Thermo Scientific Chemicals

Catalog number: 432060250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium chloride hydrate, 96%, Thermo Scientific Chemicals

Catalog number: 432060250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 432060250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37451-68-6
IUPAC Name
tetrabutylazanium chloride
Molecular Formula
C16H36ClN
InChI Key
NHGXDBSUJJNIRV-UHFFFAOYSA-M
SMILES
[Cl-].CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Color)
Off-white
Water
=<10.0 % (K.F.)
Titration Argentometric
>=95.0 % (On dried substance)
Infrared spectrum
Conforms
Appearance (Form)
Low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text