4-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
4-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 432090010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36953-37-4
IUPAC Name4-bromo-1,2-dihydropyridin-2-one
Molecular FormulaC5H4BrNO
InChI KeySSLMGOKTIUIZLY-UHFFFAOYSA-N
SMILESBrC1=CC(=O)NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only