3-Quinolineboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3-Quinolineboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Quinolineboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 432110050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS171364-85-5
IUPAC Name3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular FormulaC15H18BNO2
InChI KeyARRJAONCYUAPJR-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC2=CC=CC=C2N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or solid
Melting point55°C to 60°C
Appearance (Color)White to light brown
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only