3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
Thermo Scientific Chemicals

3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D

CAS: 24493-21-8 | C6H9D4NaO2Si | 172.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4321200101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 432120010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24493-21-8
IUPAC Namesodium 3-(trimethylsilyl)(²H₄)propanoate
Molecular FormulaC6H13NaO2Si
InChI KeyOIIWPAYIXDCDNL-HGFPCDIYSA-M
SMILES[Na+].[2H]C([2H])(C([O-])=O)C([2H])([2H])[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
D-Enrichment>=97.5 %
Water=<2.0 % (Coulometric)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473077Certificate of AnalysisJun 05, 2025432120010
A0468682Certificate of AnalysisJan 25, 2025432120010
A0460409Certificate of AnalysisMar 26, 2024432120010
A0454461Certificate of AnalysisAug 25, 2023432120010
A0405628Certificate of AnalysisDec 13, 2022432120010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection